4-Iodbutylacetat

Lieferant: CHEMPUR

Synonyme: Essigsäure(4-iodbutyl)ester

FL13611.POR
CHMPFL13611.POREA 0 EUR
CHMPFL13611.POR
4-Iodbutylacetat
4-Iodbutylacetat
Formel: C₆H₁₁IO₂
Molecular Weight: 242,06 g/mol
MDL: MFCD00010664
CAS-Nummer: 40596-44-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR