6-Iod-2,4,5-trichlorphenol

Lieferant: CHEMPUR

FL13614.POR
CHMPFL13614.POREA 0 EUR
CHMPFL13614.POR
6-Iod-2,4,5-trichlorphenol
6-Iod-2,4,5-trichlorphenol
Formel: C₆H₂CI₃IO
MDL: MFCD04113610
CAS-Nummer: 35020-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR