4-Methoxy-2(5H)-furanon

Lieferant: CHEMPUR

FL13619.POR
CHMPFL13619.POREA 0 EUR
CHMPFL13619.POR
4-Methoxy-2(5H)-furanon
4-Methoxy-2(5H)-furanon
Formel: C₅H₆O₃
Molecular Weight: 114,1 g/mol
MDL: MFCD00071565
CAS-Nummer: 69556-70-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR