4-Nitro-m-anissäure
Lieferant: CHEMPUR
Synonyme:
3-Methoxy-4-nitrobenzoic acid
FL13621.POR
CHMPFL13621.POREA
0
EUR
CHMPFL13621.POR
4-Nitro-m-anissäure
4-Nitro-m-anissäure
Formel:
C₈H₇NO₅ Molecular Weight: 197,15 g/mol Siedepunkt: 405 °C (1013 hPa) Schmelzpunkt: 233 °C |
MDL:
MFCD00007353 CAS-Nummer: 5081-36-7 EINECS: 225-793-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...