5-Methoxy-2-nitrophenol

Lieferant: CHEMPUR

FL13622.POR
CHMPFL13622.POREA 0 EUR
CHMPFL13622.POR
5-Methoxy-2-nitrophenol
5-Methoxy-2-nitrophenol
Formel: C₇H₇NO₄
Molecular Weight: 169,14 g/mol
Schmelzpunkt: 95 °C
MDL: MFCD00100932
CAS-Nummer: 704-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR