1-Amino-2-methylbutan

Lieferant: CHEMPUR

Synonyme: 2-Methylbutylamin

FL13636.POR
CHMPFL13636.POREA 0 EUR
CHMPFL13636.POR
1-Amino-2-methylbutan
1-Amino-2-methylbutane
Formel: C₅H₁₃N
Molecular Weight: 87,16 g/mol
Siedepunkt: 97 °C (1013 hPa)
Dichte: 0,755 g/cm³ (18 °C)
MDL: MFCD00008147
CAS-Nummer: 96-15-1
EINECS: 202-483-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR