(±)-3-Methylcyclopentanon
Lieferant: CHEMPUR
Synonyme:
DL-3-Methylcyclopentanone
FL13643.POR
CHMPFL13643.POREA
0
EUR
CHMPFL13643.POR
(±)-3-Methylcyclopentanon
(±)-3-Methylcyclopentanon
Formel:
C₆H₁₀O Molecular Weight: 98,14 g/mol Siedepunkt: 145 °C (1013 hPa) Dichte: 0,9136 g/cm³ (0 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001416 CAS-Nummer: 1757-42-2 EINECS: 217-148-9 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...