Methyl-3-methyl-2-furoat

Lieferant: CHEMPUR

Synonyme: Methyl 3-methylfuran-2-carboxylate

FL13654.POR 013654-5G 013654-25G
CHMPFL13654.POREA 0 EUR
CHMPFL13654.POR FLUO013654-5G FLUO013654-25G
Methyl-3-methyl-2-furoat
Methyl-3-methyl-2-furoat
Formel: C₇H₈O₃
MDL: MFCD00014110
CAS-Nummer: 6141-57-7
EINECS: 228-131-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR