3-Methyl-2-nitrobenzylalkohol

Lieferant: CHEMPUR

Synonyme: 3-(Hydroxymethyl)-2-nitrotoluol

FL13658.POR
CHMPFL13658.POREA 0 EUR
CHMPFL13658.POR
3-Methyl-2-nitrobenzylalkohol
3-Methyl-2-nitrobenzylalkohol
Formel: C₈H₉NO₃
Schmelzpunkt: 45…47 °C
MDL: MFCD00007182
CAS-Nummer: 80866-76-8
EINECS: 279-579-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR