Methyl-6-chinolincarboxylat

Lieferant: CHEMPUR

FL13662.POR
CHMPFL13662.POREA 0 EUR
CHMPFL13662.POR
Methyl-6-chinolincarboxylat
Methyl-6-chinolincarboxylat
Formel: C₁₁H₉NO₂
Molecular Weight: 187,2000 g/mol
MDL: MFCD00956766
CAS-Nummer: 38896-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR