2-Methylbenzo[b]thiophen

Lieferant: CHEMPUR

FL13664.POR
CHMPFL13664.POREA 0 EUR
CHMPFL13664.POR
2-Methylbenzo[b]thiophen
2-Methylbenzo[b]thiophen
Formel: C₉H₈S
Molecular Weight: 148,23 g/mol
MDL: MFCD00216250
CAS-Nummer: 1195-14-8
EINECS: 214-792-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR