2-Methoxy-4-nitrophenol

Lieferant: CHEMPUR

FL13672.POR
CHMPFL13672.POREA 0 EUR
CHMPFL13672.POR
2-Methoxy-4-nitrophenol
2-Methoxy-4-nitrophenol
Formel: C₇H₇NO₄
Schmelzpunkt: 103…104 °C
MDL: MFCD00012143
CAS-Nummer: 3251-56-7
EINECS: 221-839-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR