2-Nitrophenyloctylether

Lieferant: CHEMPUR

Synonyme: 1-Nitro-2-(n-octyloxy)benzene, 1-Nitro-2-(octyloxy)benzene, 2-(n-Octyloxy)nitrobenzene

FL13676.POR
CHMPFL13676.POREA 0 EUR
CHMPFL13676.POR
2-Nitrophenyloctylether
2-Nitrophenyloctylether
Formel: C₁₄H₂₁NO₃
Molecular Weight: 251,32 g/mol
MDL: MFCD00014693
CAS-Nummer: 37682-29-4
EINECS: 253-623-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR