4-Nitrophenyloctylether

Lieferant: CHEMPUR

FL13677.POR
CHMPFL13677.POREA 0 EUR
CHMPFL13677.POR
4-Nitrophenyloctylether
4-Nitrophenyloctylether
Formel: C₁₄H₂₁NO₃
MDL: MFCD00043611
CAS-Nummer: 49562-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR