6-Nitro-2,4,5-trichlorphenol

Lieferant: CHEMPUR

FL13679.POR
CHMPFL13679.POREA 0 EUR
CHMPFL13679.POR
6-Nitro-2,4,5-trichlorphenol
6-Nitro-2,4,5-trichlorphenol
Formel: C₆H₂Cl₃NO₃
MDL: MFCD01707216
CAS-Nummer: 82-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR