1,3-Phenylendiisocyanat

Lieferant: CHEMPUR

FL13690.POR
CHMPFL13690.POREA 0 EUR
CHMPFL13690.POR
1,3-Phenylendiisocyanat
1,3-Phenylendiisocyanat
Formel: C₈H₄N₂O₂
MDL: MFCD00002018
CAS-Nummer: 123-61-5
EINECS: 204-637-7
UN: 2811
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR