1-(1-Cyclopenten-1-yl)pyrrolidin

Lieferant: CHEMPUR

FL13704.POR
CHMPFL13704.POREA 0 EUR
CHMPFL13704.POR
1-(1-Cyclopenten-1-yl)pyrrolidin
1-(1-Cyclopenten-1-yl)pyrrolidin
Formel: C₉H₁₅N
MDL: MFCD00003162
CAS-Nummer: 7148-07-4
EINECS: 230-463-6
UN: 1993
ADR: 3,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR