2,2':5',2''-Terthiophen

Lieferant: CHEMPUR

FL13708.POR
CHMPFL13708.POREA 0 EUR
CHMPFL13708.POR
2,2':5',2''-Terthiophen
2,2':5',2''-Terthiophen
Formel: C₁₂H₈S₃
Molecular Weight: 248,39 g/mol
MDL: MFCD00012167
CAS-Nummer: 1081-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR