Tetrabrom-p-benzochinon

Lieferant: CHEMPUR

Synonyme: 4-Bromanil, Tetrabromchinon, p-Bromanil, 2,3,5,6-Tetrabrom-1,4-benzochinon, Tetrabrom-1,4-benzochinon

FL13709.POR
CHMPFL13709.POREA 0 EUR
CHMPFL13709.POR
Tetrabrom-p-benzochinon
Tetrabrom-p-benzochinon
Formel: C₆Br₄O₂
Molecular Weight: 423,68 g/mol
MDL: MFCD00013785
CAS-Nummer: 488-48-2
EINECS: 207-679-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR