Benzo[b]thiophen-2-carbaldehyd

Lieferant: CHEMPUR

Synonyme: Benzo[b]thiophene-2-carbaldehyde

FL13717.POR
CHMPFL13717.POREA 0 EUR
CHMPFL13717.POR
Benzo[b]thiophen-2-carbaldehyd
Benzo[b]thiophen-2-carbaldehyd
Formel: C₉H₆OS
Molecular Weight: 162,21 g/mol
MDL: MFCD01075041
CAS-Nummer: 3541-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR