2-Benzo[b]thiophencarbonsäure

Lieferant: CHEMPUR

FL13718.POR
CHMPFL13718.POREA 0 EUR
CHMPFL13718.POR
2-Benzo[b]thiophencarbonsäure
2-Benzo[b]thiophencarbonsäure
Formel: C₉H₆O₂S
Molecular Weight: 178,22 g/mol
MDL: MFCD00051636
CAS-Nummer: 6314-28-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR