4-(2-Thienyl)buttersäure

Lieferant: CHEMPUR

Synonyme: 4-(Thiophen-2-yl)butanoic acid

FL13720.POR
CHMPFL13720.POREA 0 EUR
CHMPFL13720.POR
4-(2-Thienyl)buttersäure
4-(2-Thienyl)buttersäure
Formel: C₈H₁₀O₂S
Molecular Weight: 170,23 g/mol
MDL: MFCD00005463
CAS-Nummer: 4653-11-6
EINECS: 225-090-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR