2,5-Thiophendicarbonsäure

Lieferant: CHEMPUR

Synonyme: Thiophene-2,5-dicarboxylic acid

FL13724.POR
CHMPFL13724.POREA 0 EUR
CHMPFL13724.POR
2,5-Thiophendicarbonsäure
2,5-Thiophenedicarboxylic acid
Formel: C₆H₄O₄S
Molecular Weight: 172,16 g/mol
MDL: MFCD00016896
CAS-Nummer: 4282-31-9
EINECS: 224-284-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR