4-Thioxo-3,4-dihydro-2(1H)-pyrimidinon

Lieferant: CHEMPUR

Synonyme: 2-Hydroxy-4-mercaptopyrimidin

FL13726.POR
CHMPFL13726.POREA 0 EUR
CHMPFL13726.POR
4-Thioxo-3,4-dihydro-2(1H)-pyrimidinon
4-Thiouracil
Formel: C₄H₄N₂OS
Molecular Weight: 128,15 g/mol
MDL: MFCD00090842
CAS-Nummer: 591-28-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR