α,α,α-Trifluor-o-tolylisocyanat

Lieferant: CHEMPUR
Achtung

Synonyme: 2-(Trifluormethylphenyl)isocyanat

FL13733.POR
CHMPFL13733.POREA 0 EUR
CHMPFL13733.POR
α,α,α-Trifluor-o-tolylisocyanat
α,α,α-Trifluor-o-tolylisocyanat
Formel: C₈H₄F₃NO
Molecular Weight: 187,12 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00002008
CAS-Nummer: 2285-12-3
EINECS: 218-925-5
UN: 2285
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR