9-Undecin-1-ol

Lieferant: CHEMPUR

FL13740.POR
CHMPFL13740.POREA 0 EUR
CHMPFL13740.POR
9-Undecin-1-ol
9-Undecin-1-ol
Formel: C₁₁H₂₀O
MDL: MFCD04116376
CAS-Nummer: 177961-61-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR