10-Undecinsäure

Lieferant: CHEMPUR

Synonyme: Undec-10-ynoic acid

FL13742.POR
CHMPFL13742.POREA 0 EUR
CHMPFL13742.POR
10-Undecinsäure
10-Undecynoic acid
Formel: C₁₁H₁₈O₂
Molecular Weight: 182,26 g/mol
MDL: MFCD00014389
CAS-Nummer: 2777-65-3
EINECS: 220-471-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR