4-Brom-3-nitroanilin

Lieferant: CHEMPUR

FL13743.POR
CHMPFL13743.POREA 0 EUR
CHMPFL13743.POR
4-Brom-3-nitroanilin
4-Brom-3-nitroanilin
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217,02 g/mol
MDL: MFCD01320678
CAS-Nummer: 53324-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR