5-Methoxyanthranilsäure

Lieferant: CHEMPUR
Achtung

FL13749.POR
CHMPFL13749.POREA 0 EUR
CHMPFL13749.POR
5-Methoxyanthranilsäure
5-Methoxyanthranilsäure
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016509
CAS-Nummer: 6705-03-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR