3-Amino-2,4,5-trifluorbenzoesäure

Lieferant: CHEMPUR

FL13754.POR
CHMPFL13754.POREA 0 EUR
CHMPFL13754.POR
3-Amino-2,4,5-trifluorbenzoesäure
3-Amino-2,4,5-trifluorobenzoic acid
Formel: C₇H₄F₃NO₂
MDL: MFCD00077453
CAS-Nummer: 119385-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR