2',5'-Bis(trifluormethyl)acetophenon

Lieferant: CHEMPUR

Synonyme: 1-[2,5-Bis(trifluormethyl)phenyl]ethan-1-on

FL13756.POR
CHMPFL13756.POREA 0 EUR
CHMPFL13756.POR
2',5'-Bis(trifluormethyl)acetophenon
2',5'-Bis(trifluormethyl)acetophenon
Formel: C₁₀H₆F₆O
Molecular Weight: 256,15 g/mol
MDL: MFCD04116044
CAS-Nummer: 545410-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR