2,5-Bis(trifluormethyl)benzoylchlorid

Lieferant: CHEMPUR

FL13761.POR 013761-25G 013761-5G
CHMPFL13761.POREA 0 EUR
CHMPFL13761.POR FLUO013761-25G FLUO013761-5G
2,5-Bis(trifluormethyl)benzoylchlorid
2,5-Bis(trifluormethyl)benzoylchlorid
Formel: C₉H₃ClF₆O
MDL: MFCD00074997
CAS-Nummer: 393-82-8
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR