2,5-Bis(trifluormethyl)benzylalkohol

Lieferant: CHEMPUR

FL13762.POR
CHMPFL13762.POREA 0 EUR
CHMPFL13762.POR
2,5-Bis(trifluormethyl)benzylalkohol
2,5-Bis(trifluormethyl)benzylalkohol
Formel: C₉H₆F₆O
Molecular Weight: 244,14 g/mol
MDL: MFCD00799484
CAS-Nummer: 302911-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR