2,5-Bis(trifluormethyl)benzoylchlorid

Lieferant: CHEMPUR

Synonyme: 2,5-Bis(trifluormethyl)benzoesäurechlorid

FL13765.POR
CHMPFL13765.POREA 0 EUR
CHMPFL13765.POR
2,5-Bis(trifluormethyl)benzoylchlorid
2,5-Bis(trifluormethyl)benzoylchlorid
Formel: C₉H₃ClF₆O
Molecular Weight: 276.57 g/mol
MDL: MFCD00074997
CAS-Nummer: 393-82-8
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR