(2,5-Bis(trifluormethyl)phenyl)essigsäure

Lieferant: CHEMPUR
Achtung

FL13766.POR
CHMPFL13766.POREA 0 EUR
CHMPFL13766.POR
(2,5-Bis(trifluormethyl)phenyl)essigsäure
(2,5-Bis(trifluormethyl)phenyl)essigsäure
Formel: (C₆H₃(CF₃)₂)OCH₂COOH
Molecular Weight: 272,15 g/mol
CAS-Nummer: 302912-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR