4-Brom-3,5-bis(trifluormethyl)anilin

Lieferant: CHEMPUR

FL13771.POR
CHMPFL13771.POREA 0 EUR
CHMPFL13771.POR
4-Brom-3,5-bis(trifluormethyl)anilin
4-Brom-3,5-bis(trifluormethyl)anilin
Formel: C₈H₄BrF₆N
Molecular Weight: 308,0185 g/mol
MDL: MFCD04973750
CAS-Nummer: 268733-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR