4-Brom-2,6-difluorbenzonitril
Lieferant: CHEMPUR
FL13773.POR
013773-1G
013773-5G
CHMPFL13773.POREA
0
EUR
CHMPFL13773.POR
FLUO013773-1G
FLUO013773-5G
4-Brom-2,6-difluorbenzonitril
4-Brom-2,6-difluorbenzonitril
Formel:
C₇H₂BrF₂N |
MDL:
MFCD01310981 CAS-Nummer: 123843-67-4 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...