5-Brom-2-fluorbenzamid

Lieferant: CHEMPUR

FL13777.POR 013777-1G
CHMPFL13777.POREA 0 EUR
CHMPFL13777.POR FLUO013777-1G
5-Brom-2-fluorbenzamid
5-Brom-2-fluorbenzamid
Formel: C₇H₅BrFNO
Molecular Weight: 218.02 g/mol
MDL: MFCD03094430
CAS-Nummer: 214210-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR