4-Brom-5-fluor-2-nitrotoluol

Lieferant: CHEMPUR

FL13783.POR
CHMPFL13783.POREA 0 EUR
CHMPFL13783.POR
4-Brom-5-fluor-2-nitrotoluol
4-Brom-5-fluor-2-nitrotoluol
Formel: C₇H₅BrFNO₂
Molecular Weight: 234,02 g/mol
MDL: MFCD04039299
CAS-Nummer: 224185-19-7
EINECS: 262-536-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR