2-Brom-4-(trifluormethoxy)anilin

Lieferant: CHEMPUR

Synonyme: 2-Bromo-4-trifluoromethoxyaniline

FL13789.POR
CHMPFL13789.POREA 0 EUR
CHMPFL13789.POR
2-Brom-4-(trifluormethoxy)anilin
2-Brom-4-(trifluormethoxy)anilin
Formel: C₇H₅BrF₃NO
Molecular Weight: 256,02 g/mol
MDL: MFCD00203478
CAS-Nummer: 175278-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR