1-Chlor-2,6-difluorbenzol

Lieferant: CHEMPUR

Synonyme: 2-Chloro-1,3-difluorobenzene

FL13795.POR
CHMPFL13795.POREA 0 EUR
CHMPFL13795.POR
1-Chlor-2,6-difluorbenzol
1-Chloro-2,6-difluorobenzene
Formel: C₆H₃ClF₂
Molecular Weight: 148,5378 g/mol
MDL: MFCD00142604
CAS-Nummer: 38361-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR