2-Chlor-4-fluorphenylacetonitril

Lieferant: CHEMPUR

Synonyme: 2-(2-Chloro-4-fluorophenyl)acetonitrile

FL13802.POR
CHMPFL13802.POREA 0 EUR
CHMPFL13802.POR
2-Chlor-4-fluorphenylacetonitril
2-Chlor-4-fluorphenylacetonitril
Formel: C₈H₅ClFN
MDL: MFCD00236029
CAS-Nummer: 75279-56-0
UN: 3439
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR