3-Chlor-4-fluorphenylboronsäure

Lieferant: CHEMPUR

Synonyme: Dihydroxy(3-chlor-4-fluorphenyl)boran, (3-Chlor-4-fluorphenyl)metaborsäure, 3-Chlor-4-fluorbenzolboronsäure

FL13803.POR
CHMPFL13803.POREA 0 EUR
CHMPFL13803.POR
3-Chlor-4-fluorphenylboronsäure
3-Chlor-4-fluorphenylboronsäure
Formel: C₆H₅BClFO₂
Molecular Weight: 174,37 g/mol
MDL: MFCD00051800
CAS-Nummer: 144432-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR