2,3-Difluor-4-ethoxybenzonitril

Lieferant: CHEMPUR

Synonyme: 4-Ethoxy-2,3-difluorobenzonitrile

FL13814.POR
CHMPFL13814.POREA 0 EUR
CHMPFL13814.POR
2,3-Difluor-4-ethoxybenzonitril
2,3-Difluor-4-ethoxybenzonitril
Formel: C₉H₇F₂NO
Molecular Weight: 183,1548 g/mol
MDL: MFCD04973770
CAS-Nummer: 126162-96-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR