2,3-Difluor-4-methoxybenzoesäure

Lieferant: CHEMPUR

FL13818.POR
CHMPFL13818.POREA 0 EUR
CHMPFL13818.POR
2,3-Difluor-4-methoxybenzoesäure
2,3-Difluoro-4-methoxybenzoic acid
Formel: C₈H₆F₂O₃
MDL: MFCD03428569
CAS-Nummer: 329014-60-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR