2,3-Difluor-4-methoxybenzonitril

Lieferant: CHEMPUR

Synonyme: 2,3-Difluor-4-methoxybenzoesäurenitril

FL13820.POR
CHMPFL13820.POREA 0 EUR
CHMPFL13820.POR
2,3-Difluor-4-methoxybenzonitril
2,3-Difluor-4-methoxybenzonitril
Formel: C₈H₅F₂NO
Molecular Weight: 169,13 g/mol
MDL: MFCD03428596
CAS-Nummer: 256417-12-6
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR