3,4-Difluor-5-(trifluormethyl)benzoesäure

Lieferant: CHEMPUR

FL13828.POR
CHMPFL13828.POREA 0 EUR
CHMPFL13828.POR
3,4-Difluor-5-(trifluormethyl)benzoesäure
3,4-Difluoro-5-(trifluoromethyl)benzoic acid
Formel: C₈H₃F₅O₂
MDL: MFCD00236257
CAS-Nummer: 237761-76-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR