3-Fluor-4-hydroxybenzonitril
Lieferant: CHEMPUR
Synonyme:
3-Fluor-4-hydroxybenzoesäurenitril
FL13830.POR
013830-25G
013830-5G
CHMPFL13830.POREA
0
EUR
CHMPFL13830.POR
FLUO013830-25G
FLUO013830-5G
3-Fluor-4-hydroxybenzonitril
3-Fluor-4-hydroxybenzonitril
Formel:
C₇H₄FNO Molecular Weight: 137.11 g/mol |
MDL:
MFCD00215834 CAS-Nummer: 405-04-9 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...