4-Fluor-o-anisaldehyd

Lieferant: CHEMPUR

Synonyme: 4-Fluor-2-anisaldehyd, 4-Fluor-2-methoxybenzaldehyd

FL13832.POR
CHMPFL13832.POREA 0 EUR
CHMPFL13832.POR
4-Fluor-o-anisaldehyd
4-Fluor-o-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
MDL: MFCD00143318
CAS-Nummer: 450-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR