4-Fluor-2-methoxybenzoesäure

Lieferant: CHEMPUR

FL13833.POR
CHMPFL13833.POREA 0 EUR
CHMPFL13833.POR
4-Fluor-2-methoxybenzoesäure
4-Fluoro-2-methoxybenzoic acid
Formel: C₈H₇FO₃
MDL: MFCD00673010
CAS-Nummer: 395-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR